Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPA™ ChemZoil™ NL339 Chemical Resistant Neoprene Gloves

Catalog No. 19046579 Shop All MAPA Professional Products
Change view
Click to view available options
Size:
8
9
10
11
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-046-579 8
19-046-580 9
19-046-581 10
19-046-582 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-046-579 Supplier MAPA™ Supplier No. 339428
Add to Cart
Add to Cart
This item is not returnable. View return policy

Neoprene multi-layer gloves provide chemical resistance against acids and alkalis, such as concentrated hydrofluoric acid, oils, and greases.

  • Protects against acids
  • For use with chemical protective suits
  • Black, heavyweight, neoprene
  • Articulated finger design for added comfort, flexibility and reduced hand fatigue
  • Soft, interlocked knit cotton liner absorbs perspiration and adds comfort
  • Multilayer construction assures a leakproof glove
  • Roughened grip with pinked cuff
  • Length: 14 in. (35.6 cm)

Specifications

Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) Cat III CE 0334, EN 407:X1XXXX, EN 511
Color Black
Latex Free Yes
Material Neoprene
Size 8
Thickness (English) 50 mil
Thickness (Metric) 1.20 mm
EN388 Standard EN 388:3121
EN 421 Yes
EN ISO 374-1 CHEMICAL LETTER CODES ABCJKL
EN ISO 374 - 5 Bacteria, Fungi only
For Use With (Application) Chemical processing, alkaline units at petroleum refineries, metal plating operations, hazardous material handling, hazmat and chemical spills
Glove Cuff Style Pinked
Grip Design Pebbled
Liner Material Cotton Knit
Lining Lined
Model 339428
Series ChemZoil™ NL-339
Length (English) 14 in.
Length (Metric) 35.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.