Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPA™ Cobra™ FluoroSolv™ Gloves

Catalog No. 19999346 Shop All MAPA Professional Products
Change view
Click to view available options
Size:
8
9
10
11
Length (English):
11 in.
14 in.
Length (Metric):
28 cm
35.5 cm
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Length (English) Length (Metric)
19-999-346 9 14 in. 35.5 cm
19-050-581A 8 11 in. 28 cm
19-050-581B 9 11 in. 28 cm
19-050-581C 10 11 in. 28 cm
19-050-581D 11 11 in. 28 cm
19-999-345 8 14 in. 35.5 cm
19-999-343 10 14 in. 35.5 cm
19-999-344 11 14 in. 35.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-999-346 Supplier MAPA™ Supplier No. 467439

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Resist aromatic solvents

Unsupported powder-free fluoroelastomer/nitrile gloves resist aromatic solvents, chloroform, and PCBs. Suitable for use in refineries, plating operations, laboratories, and hazardous material handling.

  • Consist of a layer of black FluoroSolv fluoroelastomer polymer over a green, powder-free nitrile shell
  • Two-tone layering serves as a visual wear indicator
  • Nitrile layer also provides increased cut, puncture, and tear resistance
  • Z-pattern texture channels chemicals away from glove surface, improving wet and dry grip
  • Hand-specific and unlined with a flat cuff
  • Embossed with size measurement for easy identification
  • Style F-466 is 11 in. long (28 cm), 15 mil thick
  • Style F-467 is 14 in. long (36 cm), 25 mil thick

Not for use with ketonic solvents.

Specifications

Powdered Powder Free
Size 9
Length (English) 14 in.
Length (Metric) 35.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.