Learn More
MSC MAPA Professional Cut, Puncture and Abrasion-Resistant Gloves, 1 Pair
Catalog No. 5000792017
Encompass
These gloves are designed for cut, puncture, and abrasion resistance. Featuring a polyurethane coating on the palm and fingertips, they provide a rough grip and are suitable for various industrial applications.
  • Knit style
  • Polyurethane coating on palm and fingertips for rough grip
  • Designed for cut, puncture, and abrasion resistance
  • ANSI/ISEA cut resistance level A2
  • ANSI/ISEA puncture resistance level 2
  • ANSI/ISEA abrasion resistance level 4
  • Gauge: 13, Size: 2X-Large
  • EN 388 rating 4342BX
  • High-density polyethylene (HDPE) seamless lining
  • Excellent dexterity from precise fit
  • Ergonomic design reduces hand fatigue
  • Washable up to 5 times at 40°C for extended lifespan and reduced waste
  • Silicone-free to prevent defects on metal and glass surfaces
  • Unisex design
Catalog No. 50-007-92017 Supplier MSC Supplier No. 65722803
These gloves are designed for cut, puncture, and abrasion resistance. Featuring a polyurethane coating on the palm and fingertips, they provide a rough grip and are suitable for various industrial applications.
  • Knit style
  • Polyurethane coating on palm and fingertips for rough grip
  • Designed for cut, puncture, and abrasion resistance
  • ANSI/ISEA cut resistance level A2
  • ANSI/ISEA puncture resistance level 2
  • ANSI/ISEA abrasion resistance level 4
  • Gauge: 13, Size: 2X-Large
  • EN 388 rating 4342BX
  • High-density polyethylene (HDPE) seamless lining
  • Excellent dexterity from precise fit
  • Ergonomic design reduces hand fatigue
  • Washable up to 5 times at 40°C for extended lifespan and reduced waste
  • Silicone-free to prevent defects on metal and glass surfaces
  • Unisex design