Learn More
MSC MAPA Professional Gloves, Trilites 994, Nitrile/Latex/Polychloroprene Blend, 6.0 mil, 100/Pk.
Catalog No. 5000795430
Encompass
The Trilites 994 Series disposable gloves are designed for general purpose applications. These gloves are made from a blend of Nitrile, Latex, and Polychloroprene, offering chemical splash protection.
  • Made from a Nitrile, Latex and Polychloroprene blend
  • Thickness of 6.0 mil
  • Pebbled (full) grip surface
  • General purpose grade
  • Extended cuff type
  • Length is 10.0 in.
  • Medium glove size
  • Mauve color
  • Acceptable Quality Level (AQL) of 1.5
  • Non-sterile
  • Powder-free
  • Rolled cuff style
  • Pebbled texture
  • Ambidextrous
  • Meets standards CAT III 0493; EN ISO 374-5; EN ISO 374-1:2016/Type B
  • ISO 9001 rated
  • Provides chemical splash protection
Catalog No. 50-007-95430 Supplier MSC Supplier No. 07725724
The Trilites 994 Series disposable gloves are designed for general purpose applications. These gloves are made from a blend of Nitrile, Latex, and Polychloroprene, offering chemical splash protection.
  • Made from a Nitrile, Latex and Polychloroprene blend
  • Thickness of 6.0 mil
  • Pebbled (full) grip surface
  • General purpose grade
  • Extended cuff type
  • Length is 10.0 in.
  • Medium glove size
  • Mauve color
  • Acceptable Quality Level (AQL) of 1.5
  • Non-sterile
  • Powder-free
  • Rolled cuff style
  • Pebbled texture
  • Ambidextrous
  • Meets standards CAT III 0493; EN ISO 374-5; EN ISO 374-1:2016/Type B
  • ISO 9001 rated
  • Provides chemical splash protection