Learn More
MSC MAPA Professional Gloves, Trilites, X-Large, 6 mil, Powder-Free, 1 Case
Catalog No. 5000803235
Encompass
These gloves are constructed from a unique TRI-polymer blend of neoprene, nitrile, and natural rubber, offering the comfort of natural rubber with enhanced protection. They are unlined, powder-free, and ambidextrous.
  • Unique TRI-polymer blend for comfort and protection
  • Unlined for direct contact feel
  • Powder-free to minimize contamination
  • Ambidextrous design for ease of use
  • Textured grip for enhanced handling
  • Rolled cuff for added durability
  • Non-sterile for general laboratory applications
Catalog No. 50-008-03235 Supplier MSC Supplier No. 08405326
These gloves are constructed from a unique TRI-polymer blend of neoprene, nitrile, and natural rubber, offering the comfort of natural rubber with enhanced protection. They are unlined, powder-free, and ambidextrous.
  • Unique TRI-polymer blend for comfort and protection
  • Unlined for direct contact feel
  • Powder-free to minimize contamination
  • Ambidextrous design for ease of use
  • Textured grip for enhanced handling
  • Rolled cuff for added durability
  • Non-sterile for general laboratory applications