Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPA™ Krynit Grip and Proof 580 Cut-Resistant Gloves, Nitrile Coated
GREENER_CHOICE

Catalog No. 19249825 Shop All MAPA Professional Products
Change view
Click to view available options
Size:
7
8
9
10
11
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-249-825 11
19-249-821 7
19-249-822 8
19-249-823 9
19-249-824 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-249-825 Supplier MAPA™ Supplier No. 34580031
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Grip and proof nitrile coating on the palm and fingers ensures a secure grip in oily environments. Oil-proof gloves are ideal for jobs such as handling sheet metal, working with oily mechanical parts or sorting small, sharp parts.

  • Nitrile coating on the palm and fingers ensures secure grip in oily environments
  • Ideal for handling sheet metal, working with oily mechanical parts and sorting small and sharp parts
  • HDPE provides cut and abrasion resistance
  • CE level 3 cut resistance/ANSI A2
  • Medium weight

Specifications

Certifications/Compliance (Europe) CE
Coating Material Nitrile
Color Black
Cut Resistance Rating ANSI A2
Material Nitrile
Product Type Cut-resistant Gloves
Size 11
For Use With (Application) Handling sheet metal, working with oily mechanical parts, sorting small/sharp parts
Glove Cuff Style Knit
Model 580411
Series Krynit Grip & Proof 580
Length (English) 9 to 10 in.
Length (Metric) 22.8 to 25.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.