Learn More
Cole-Parmer MAPA 332420 Heat-Insulated Neoprene Gloves, Extra-large
Catalog No. NC0503616 Shop All Cole-Parmer Products
Encompass_Preferred
These gloves are made of synthetic neoprene rubber for good chemical resistance. Easily maintain your grip with the nonslip, roughened surface. Withstand temperatures from -100 to 480°F (-73 to 249°C). The 14"L gloves feature integral cotton fleece liner.
This item is not returnable. View return policy
Catalog No. NC0503616 Supplier Cole-Parmer Supplier No. 4021132
These gloves are made of synthetic neoprene rubber for good chemical resistance. Easily maintain your grip with the nonslip, roughened surface. Withstand temperatures from -100 to 480°F (-73 to 249°C). The 14"L gloves feature integral cotton fleece liner.