Learn More
Cayman Chemical MapenterolhydrochlorIde 10mg
SDP
Catalog No. 502947909
Encompass_Preferred
A β2-AR agonist; has been used as a standard for the simultaneous detection of multiple drug residues in commercial livestock synovial fluids by GC-MS; has been used as an illicit feed additive to promote growth in livestock
Catalog No. 50-294-7909 Supplier Cayman Chemical Supplier No. 3456210 MG
A β2-AR agonist; has been used as a standard for the simultaneous detection of multiple drug residues in commercial livestock synovial fluids by GC-MS; has been used as an illicit feed additive to promote growth in livestock