Learn More
Selleck Chemical LLC MAPK Inhibitor Library 100uL/well
SDP
Catalog No. 501948202
Encompass_Preferred
A unique collection of 259 small molecule inhibitors used for MAPK signaling research. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8202 Supplier Selleck Chemical LLC Supplier No. L3400100UL/WELL
Add to Cart
Add to Cart
Request a Quote
A unique collection of 259 small molecule inhibitors used for MAPK signaling research. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.