Learn More
Aviva Systems Biology Corp MAPK1 Antibody - middle region (ARP58900_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58900_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAPK1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human MAPK1. Alternate names: ERK, ERK2, ERT1, MAPK2, P42MAPK, PRKM1, PRKM2, p38, p40, p41, p41mapkApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-522