Learn More
Aviva Systems Biology Corp Mapk10 Antibody - N-terminal region (ARP57728_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57728_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Mapk10. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Jnk3, SAPb, SAPKC, Serk2ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-366