Learn More
Aviva Systems Biology Corp Mapk12 Antibody - C-terminal region (ARP56536_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56536_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Mapk12. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AW123708, Erk6, P38gamma, Prkm12, Sapk3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-174