Learn More
Aviva Systems Biology Corp MAPK12 Antibody - N-terminal region (ARP56535_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56535_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAPK12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MAPK12. Alternate names: ERK3, ERK6, P38GAMMA, PRKM12, SAPK-3, SAPK3ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-61895