Learn More
Aviva Systems Biology Corp MAPK13 Antibody - middle region (ARP56439_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56439_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAPK13. It was validated on Western Blot. MAPK13,mitogen-activated protein kinase 13,SAPK4; PRKM13; MGC99536; p38delta; MAPK13,mitogen-activated protein kinase 13,mitogen-activated protein kinase 13,MAPK 13,MAP kinase 13,MAP kinase p38 delta,stress-activated protein kinase 4,mitogen-activated protein kinase p38 deltaThe immunogen is a synthetic peptide directed towards the middle region of human MAPK13. Alternate names: MGC99536, PRKM13, SAPK4, p38deltaApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-286-153