Learn More
Aviva Systems Biology Corp MAPK13 Antibody - middle region (ARP56440_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56440_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAPK13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MAPK13. Alternate names: MGC99536, PRKM13, SAPK4, p38deltaApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-61861