Learn More
Medchemexpress LLC HY-18850 5mg Medchemexpress, MAPK13-IN-1 CAS:229002-10-2 Purity:>98%
SDP
Catalog No. 501970015
Encompass_Preferred
Medchemexpress, HY-18850 5mg MAPK13-IN-1 CAS:229002-10-2 MPAK13-IN-1 is a MAPK13 (p38δ) inhibitor, with an IC50 of 620 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0015 Supplier Medchemexpress LLC Supplier No. HY188505MG
Medchemexpress, HY-18850 5mg MAPK13-IN-1 CAS:229002-10-2 MPAK13-IN-1 is a MAPK13 (p38δ) inhibitor, with an IC50 of 620 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.