Learn More
Cayman Chemical MAPK13 Inhibitor Compound 61, Purity: ≥98%, CAS: 229002-10-2, 5 mg
SDP
Catalog No. 502944945
Encompass_Preferred
A p38δ MAPK inhibitor (IC50 = 620 nM)
Catalog No. 50-294-4945 Supplier Cayman Chemical Supplier No. 421215 MG
A p38δ MAPK inhibitor (IC50 = 620 nM)