Learn More
Aviva Systems Biology Corp MAPK4 Antibody - middle region (ARP56433_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56433_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAPK4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MAPK4. Alternate names: ERK3, Erk4, PRKM4, p63MAPKApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61860