Learn More
Aviva Systems Biology Corp MAPK6 Antibody - middle region (ARP56436_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56436_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAPK6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human MAPK6. Alternate names: DKFZp686F03189, ERK3, HsT17250, PRKM6, p97MAPKApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-152