Learn More
Aviva Systems Biology Corp MAPK7 Antibody - middle region (ARP57731_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57731_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAPK7. It was validated on Western Blot. MAPK7,mitogen-activated protein kinase 7,BMK1; ERK4; ERK5; PRKM7; MAPK7,mitogen-activated protein kinase 7,mitogen-activated protein kinase 7,BMK-1,ERK-5,MAPK 7,BMK1 kinase,MAP kinase 7,big MAP kinase 1,extracellular signal-regulated kinase 5,extracellular-signal-regulated kinase 5The immunogen is a synthetic peptide directed towards the middle region of human MAPK7. Alternate names: BMK1, ERK4, ERK5, PRKM7ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-367