Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MAPK9 (JNK2), His Tag Recombinant Protein

Catalog No. PR6568B
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR6568B 100 μg
PV3620 10 μg
PV4018 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PR6568B Supplier Invitrogen™ Supplier No. PR6568B
Only null left
Add to Cart
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

MAPK9 (JNK2) is a serine/threonine kinase which plays an important role in the MAPK pathway. JNK2 (p54a, SAPK1a), along with JNK1 and JNK3, is thought to play an important role in nuclear signal transduction through its environmental stress activation and subsequent phosphorylation of the nuclear transcription factor p53.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P45984
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 0.02% Triton X-100, 50% glycerol, 0.5mM EDTA, 150mM NaCl, 2mM DTT and no preservative; pH 7.5
Gene ID (Entrez) 5601
Molecular Weight (g/mol) 52.2 kDa
Name Human MAPK9 (JNK2), His Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Source Human
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AI851083; c-Jun kinase 2; c-Jun N-terminal kinase 2; I79_012561; JNK; JNK/SAPK alpha; JNK1; JNK1A2; JNK2; JNK21B1/2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; JNK-46; JNK-55; Jun kinase; MAP kinase 10; MAP kinase 8; MAP kinase 9; MAPK 10; MAPK 9; MAPK10; MAPK8; Mapk9; mitogen activated protein kinase 9; mitogen-activated protein kinase 9; OTTHUMP00000161542; OTTHUMP00000161543; p54a; p54-alpha; p54aSAPK; PRKM8; PRKM9; protein kinase, mitogen-activated 9; SAPK; SAPK1; SAPK1a; SAPK1c; SAPK-alpha; stress activated protein kinase alpha II; stress-activated protein kinase 1a; Stress-activated protein kinase JNK2
Common Name JNK2
Gene Symbol MAPK9
Kinase Family MAP Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Ser⁄Thr Kinases: CMGC Group
Shipping Condition Dry Ice
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.