Learn More
Eurofins Discoverx MAPKAP Kinase 3, active, 10 µg. (14-585)
Supplier:  Eurofins Discoverx 14585
Encompass
N-terminal GST-tagged recombinant, human MAPKAP Kinase 3 amino acids 2-end; activated using SAPK2a (Catalog #14-587). For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9009