Learn More
Aviva Systems Biology Corp MAPKAPK2 Antibody - middle region (ARP60320_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60320_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAPKAPK2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human MAPKAPK2. Alternate names: 3PK, MAPKAP3, MK2, MK-2, MAPKAP-K2ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-004