Learn More
Enzo Life Sciences MAPKAPK2 substrate (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP1960001
Encompass_Preferred
This peptide, KKLNRTLSVA is derived from the N-terminus of glycogen synthase and is known to be a good substrate for MAPKAPK2. Alternative name: MAP kinase activated protein kinase 2 substrate, Lys-Lys-Leu-Asn-Arg-Thr-Leu-Ser-Val-Ala. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1274