Learn More
Aviva Systems Biology Corp MAPKAPK3 Antibody - C-terminal region (ARP60322_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60322_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAPKAPK3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human MAPKAPK3. Alternate names: 3PK, MAPKAP3, MK-3, MAPKAP-K3, MAPKAPK-3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-006