Learn More
Sino Biological Recombinant Human MAPKAPK2/MK2 Protein (His Tag), Active
Catalog No. 502574255
Encompass_Preferred
Recombinant human MAPKAPK2 (46-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
Catalog No. 50-257-4255 Supplier Sino Biological Supplier No. M4011H50
Recombinant human MAPKAPK2 (46-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.