Learn More
Medchemexpress LLC HY-14864 10mg Medchemexpress, Mapracorat CAS:887375-26-0 Purity:>98%
SDP
Catalog No. 501932711
Encompass_Preferred
Medchemexpress, HY-14864 10mg Mapracorat CAS:887375-26-0 Mapracorat is a novel non-steroidal selective glucocorticoid receptor agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2711 Supplier Medchemexpress LLC Supplier No. HY1486410MG
Medchemexpress, HY-14864 10mg Mapracorat CAS:887375-26-0 Mapracorat is a novel non-steroidal selective glucocorticoid receptor agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.