Learn More
Medchemexpress LLC Maprotiline-d3 hydrochloride | 1329496-63-0 | 98.8% | 1 MG
SDP
Catalog No. 5000408223
Encompass_Preferred
Maprotiline-d3 hydrochloride is a deuterium labeled version of Maprotiline hydrochloride. It is intended for research use only and not for patient use. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules for use as tracers during drug development. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-08223 Supplier Medchemexpress LLC Supplier No. HYB0444S11MG
Maprotiline-d3 hydrochloride is a deuterium labeled version of Maprotiline hydrochloride. It is intended for research use only and not for patient use. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules for use as tracers during drug development. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS