Learn More
Medchemexpress LLC Maprotiline-d3 hydrochloride | 1329496-63-0 | 98.8% | 5 MG
SDP
Catalog No. 5000407747
Encompass_Preferred
Maprotiline-d3 (hydrochloride) is a deuterium labeled version of Maprotiline (hydrochloride) and is intended for research use only.
  • Can be utilized as a tracer.
  • Functions as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-07747 Supplier Medchemexpress LLC Supplier No. HYB0444S15MG
Maprotiline-d3 (hydrochloride) is a deuterium labeled version of Maprotiline (hydrochloride) and is intended for research use only.
  • Can be utilized as a tracer.
  • Functions as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs.