Learn More
Medchemexpress LLC Maprotiline-d5 hydrochloride | 1794942-12-3 | 1 MG
SDP
Catalog No. 5000407742
Encompass_Preferred
Maprotiline-d5 hydrochloride is a deuterium-labeled version of Maprotiline hydrochloride, functioning as a selective noradrenalin re-uptake inhibitor and a tetracyclic antidepressant. This compound is suitable for various quantitative analyses due to its stable heavy isotopes.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes for drug development tracing
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-07742 Supplier Medchemexpress LLC Supplier No. HYB0444S1MG
Maprotiline-d5 hydrochloride is a deuterium-labeled version of Maprotiline hydrochloride, functioning as a selective noradrenalin re-uptake inhibitor and a tetracyclic antidepressant. This compound is suitable for various quantitative analyses due to its stable heavy isotopes.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes for drug development tracing
  • Potential to affect pharmacokinetic and metabolic profiles of drugs