Learn More
Medchemexpress LLC Maprotiline (hydrochloride) | 10347-81-6 | MFCD00079464 | 100.0% | 313.86 | C20H24ClN | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0444R10MG
Encompass_Preferred
Maprotiline hydrochloride is the hydrochloride salt of maprotiline, a tetracyclic antidepressant provided as a high-purity analytical standard for research use. It is used in biochemical and pharmacological studies, including screening, assay development, and mechanistic investigations.
  • High purity (99.98%) suitable for analytical applications.
  • Hydrochloride salt; chemical formula C20H24ClN; molecular weight 313.86.
  • Intended for research, screening, and assay development.
  • Store sealed at 4°C; in solution store frozen for extended stability.
Catalog No. 50-002-97357