Learn More
Cayman Chemical Maprotiline (hydrochloride), Purity: ≥98%, CAS: 10347-81-6, 50 mg
SDP
Catalog No. 502972152
Encompass_Preferred
A tricyclic antidepressant; binds to NET (Kd = 11 nM); selective for NET over the serotonin and dopamine transporters (Kds = 5,800 and 1,000 nM, respectively); binds to 5-HT2A (Ki = 51 nM), as well as histamine H1, muscarinic acetylcholine, α1-adrenergic, and dopamine D2 receptors (Kds = 2, 570, 90, and 350 nM, respectively); inhibits norepinephrine reuptake in rat brain and peripheral tissues; reduces isolation-induced aggressive behavior and inhibits electrical foot-shock stimulation-induced belligerence in mice from 3-10 mg/kg; prevents acid sphingomyelinase activation and subsequent ceramide release induced by pp-VSV-SARS-CoV-2 infection in Vero cells at 20 µM
Catalog No. 50-297-2152 Supplier Cayman Chemical Supplier No. 1589250 MG
A tricyclic antidepressant; binds to NET (Kd = 11 nM); selective for NET over the serotonin and dopamine transporters (Kds = 5,800 and 1,000 nM, respectively); binds to 5-HT2A (Ki = 51 nM), as well as histamine H1, muscarinic acetylcholine, α1-adrenergic, and dopamine D2 receptors (Kds = 2, 570, 90, and 350 nM, respectively); inhibits norepinephrine reuptake in rat brain and peripheral tissues; reduces isolation-induced aggressive behavior and inhibits electrical foot-shock stimulation-induced belligerence in mice from 3-10 mg/kg; prevents acid sphingomyelinase activation and subsequent ceramide release induced by pp-VSV-SARS-CoV-2 infection in Vero cells at 20 µM