Learn More
Aviva Systems Biology Corp MAPT Antibody - middle region (ARP48103_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48103_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAPT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MAPT. Alternate names: DDPAC, FLJ31424, FTDP-17, MAPTL, MGC138549, MSTD, MTBT1, MTBT2, PPND, TAUApplicationIHC, WB. Species reactivity: Human, Mouse, Pig, Rat
Catalog No. 50-014-54067