Learn More
Amsbio LLC MAPTrix™ Screen for Cell Adhesion Assay, 24 different ECM mimetics coated 96 well plate (4 duplicates), 2 plates
Catalog No. 502041577
Encompass
Mussel Adhesive Protein based matrix (MAPTrix™) recombinant extracellular matrix (ECM) acts as bio-mimetic for traditional basement membrane extracts. MAPTrix™ replaces traditional ECM with genetically incorporated bioactive peptides (recognition peptides) that provide an environment for the maintenance of cells under serum and feeder-free conditions. MAPTrix™ creates the first combinatorial synthetic ECM library for engineering integrin specific surfaces.
Catalog No. 50-204-1577 Supplier Amsbio LLC Supplier No. ANN901
Mussel Adhesive Protein based matrix (MAPTrix™) recombinant extracellular matrix (ECM) acts as bio-mimetic for traditional basement membrane extracts. MAPTrix™ replaces traditional ECM with genetically incorporated bioactive peptides (recognition peptides) that provide an environment for the maintenance of cells under serum and feeder-free conditions. MAPTrix™ creates the first combinatorial synthetic ECM library for engineering integrin specific surfaces.