Learn More
Medchemexpress LLC Maralixibat chloride | 228113-66-4 | 99.6% | 710.41 | 1 MG
SDP
Catalog No. 5000391162
Encompass_Preferred
Maralixibat chloride is an orally active ileal bile acid transporter (IBAT) inhibitor. It can be used for the research of rare cholestatic liver diseases, including Alagille syndrome (ALGS), progressive familial intrahepatic cholestasis (PFIC), and biliary atresia.
  • Orally active ileal bile acid transporter (IBAT) inhibitor.
  • Can be used for research on rare cholestatic liver diseases such as Alagille syndrome (ALGS), progressive familial intrahepatic cholestasis (PFIC), and biliary atresia.
  • Reduces elevations in sBA levels, improves liver function, and reduces liver tissue damage in a rat partial bile duct ligation model of cholestasis.
Catalog No. 50-003-91162 Supplier Medchemexpress LLC Supplier No. HY167471MG
Maralixibat chloride is an orally active ileal bile acid transporter (IBAT) inhibitor. It can be used for the research of rare cholestatic liver diseases, including Alagille syndrome (ALGS), progressive familial intrahepatic cholestasis (PFIC), and biliary atresia.
  • Orally active ileal bile acid transporter (IBAT) inhibitor.
  • Can be used for research on rare cholestatic liver diseases such as Alagille syndrome (ALGS), progressive familial intrahepatic cholestasis (PFIC), and biliary atresia.
  • Reduces elevations in sBA levels, improves liver function, and reduces liver tissue damage in a rat partial bile duct ligation model of cholestasis.