Learn More
Medchemexpress LLC Maralixibat chloride | 228113-66-4 | 99.6% | 710.41 | 1 ML
SDP
Catalog No. 5000390669
Encompass_Preferred
Maralixibat chloride is an orally active ileal bile acid transporter (IBAT) inhibitor. It is used for the research of rare cholestatic liver diseases such as Alagille syndrome (ALGS), progressive familial intrahepatic cholestasis (PFIC), and biliary atresia.
  • Reduces elevated serum bile acid (sBA) levels.
  • Improves liver function.
  • Decreases liver tissue damage in rat models of cholestasis.
  • Orally active.
  • For research use only.
Catalog No. 50-003-90669 Supplier Medchemexpress LLC Supplier No. HY1674710MM/1ML
Maralixibat chloride is an orally active ileal bile acid transporter (IBAT) inhibitor. It is used for the research of rare cholestatic liver diseases such as Alagille syndrome (ALGS), progressive familial intrahepatic cholestasis (PFIC), and biliary atresia.
  • Reduces elevated serum bile acid (sBA) levels.
  • Improves liver function.
  • Decreases liver tissue damage in rat models of cholestasis.
  • Orally active.
  • For research use only.