Learn More
Medchemexpress LLC Maralixibat chloride | 228113-66-4 | 99.6% | 710.41 | 25 MG
SDP
Catalog No. 5000393925
Encompass_Preferred
Maralixibat chloride is an orally active ileal bile acid transporter (IBAT) inhibitor. It is used for the research of rare cholestatic liver diseases such as Alagille syndrome (ALGS), progressive familial intrahepatic cholestasis (PFIC), and biliary atresia. Maralixibat chloride has been shown to reduce elevations in sBA levels, improve liver function, and decrease liver tissue damage in a rat model of cholestasis.
  • Orally active ileal bile acid transporter (IBAT) inhibitor.
  • Can be used for research on rare cholestatic liver diseases, including Alagille syndrome (ALGS), progressive familial intrahepatic cholestasis (PFIC), and biliary atresia.
  • Reduces elevated sBA levels.
  • Improves liver function.
  • Reduces liver tissue damage in cholestasis models.
Catalog No. 50-003-93925 Supplier Medchemexpress LLC Supplier No. 25MGHY16747
Maralixibat chloride is an orally active ileal bile acid transporter (IBAT) inhibitor. It is used for the research of rare cholestatic liver diseases such as Alagille syndrome (ALGS), progressive familial intrahepatic cholestasis (PFIC), and biliary atresia. Maralixibat chloride has been shown to reduce elevations in sBA levels, improve liver function, and decrease liver tissue damage in a rat model of cholestasis.
  • Orally active ileal bile acid transporter (IBAT) inhibitor.
  • Can be used for research on rare cholestatic liver diseases, including Alagille syndrome (ALGS), progressive familial intrahepatic cholestasis (PFIC), and biliary atresia.
  • Reduces elevated sBA levels.
  • Improves liver function.
  • Reduces liver tissue damage in cholestasis models.