Learn More
Medchemexpress LLC Maralixibat chloride | 228113-66-4 | 99.6% | 710.41 | 5 MG
SDP
Catalog No. 5000393820
Encompass_Preferred
Maralixibat chloride (SHP625) is an orally active ileal bile acid transporter (IBAT) inhibitor. It is used for the research of rare cholestatic liver diseases including Alagille syndrome (ALGS), progressive familial intrahepatic cholestasis (PFIC), and biliary atresia. It reduces elevations in sBA levels, improves liver function, and reduces liver tissue damage in cholestasis models.
  • Orally active ileal bile acid transporter (IBAT) inhibitor
  • Suitable for research into rare cholestatic liver diseases such as Alagille syndrome, progressive familial intrahepatic cholestasis, and biliary atresia
  • Reduces elevations in sBA levels
  • Improves liver function
  • Reduces liver tissue damage in cholestasis models
Catalog No. 50-003-93820 Supplier Medchemexpress LLC Supplier No. HY167475MG
Maralixibat chloride (SHP625) is an orally active ileal bile acid transporter (IBAT) inhibitor. It is used for the research of rare cholestatic liver diseases including Alagille syndrome (ALGS), progressive familial intrahepatic cholestasis (PFIC), and biliary atresia. It reduces elevations in sBA levels, improves liver function, and reduces liver tissue damage in cholestasis models.
  • Orally active ileal bile acid transporter (IBAT) inhibitor
  • Suitable for research into rare cholestatic liver diseases such as Alagille syndrome, progressive familial intrahepatic cholestasis, and biliary atresia
  • Reduces elevations in sBA levels
  • Improves liver function
  • Reduces liver tissue damage in cholestasis models