Learn More
TARGETMOL CHEMICALS INC MARALIXIBAT CHLORIDE 5MG
SDP
Catalog No. 502580319
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Maralixibat Chloride (LUM001 chloride) an apical sodium-dependent bile acid transport inhibitor prevents enterohepatic bile acid recirculation. purity: 98%
Catalog No. 50-258-0319 Supplier TARGETMOL CHEMICALS INC Supplier No. T328735MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Maralixibat Chloride (LUM001 chloride) an apical sodium-dependent bile acid transport inhibitor prevents enterohepatic bile acid recirculation. purity: 98%