Learn More
MSC Marathon Industries Swivel Top Plate Caster, Polyurethane, 10 in, 300 lb
Catalog No. 5000531757
Encompass
A Swivel Top Plate Caster with a polyurethane wheel, featuring a 10 in wheel diameter, 3-19/64 in wheel width, 300 lb capacity, and an overall height of 12 in. It also has a 4.5 in plate length and 4 in plate width.
  • Top plate mount
  • Polyurethane wheel material
  • 10 in wheel diameter
  • 3-19/64 in wheel width
  • 300 lb load capacity
  • 12 in overall height
  • 4.5 in plate length
  • 4 x 4-1/2 in plate size
  • Swivel style
Catalog No. 50-005-31757 Supplier MSC Supplier No. 60958444
A Swivel Top Plate Caster with a polyurethane wheel, featuring a 10 in wheel diameter, 3-19/64 in wheel width, 300 lb capacity, and an overall height of 12 in. It also has a 4.5 in plate length and 4 in plate width.
  • Top plate mount
  • Polyurethane wheel material
  • 10 in wheel diameter
  • 3-19/64 in wheel width
  • 300 lb load capacity
  • 12 in overall height
  • 4.5 in plate length
  • 4 x 4-1/2 in plate size
  • Swivel style