Learn More
Apexbio Technology LLC Maraviroc(Synonyms: Selzentry, Celsentri, UK-427857, MVC), 10mM (in 1mL DMSO), CAS: 376348-65-1.
SDP
Catalog No. 5000060998
Encompass_Preferred
Maraviroc (CAS 376348-65-1) is a potent selective antagonist of the chemokine receptor CCR5 CCR5 functions as a coreceptor expressed on immune cells and facilitates entry of certain HIV-1 virus strains (R5-tropic) By binding CCR5 maraviroc effectively blocks HIV-1 envelope protein gp120 anchoring and consequently prevents viral fusion and subsequent cell entry In cellular assays with CCR5-tropic HIV-1 maraviroc demonstrates substantial antiviral activity (IC50 approximately 2 0 nM) and efficiently inhibits gp120-CCR5 interaction Maraviroc serves as a valuable research tool to explore CCR5-mediated viral entry and HIV tropism dynamics
Catalog No. 50-000-60998 Supplier Apexbio Technology LLC Supplier No. A83115.1
Maraviroc (CAS 376348-65-1) is a potent selective antagonist of the chemokine receptor CCR5 CCR5 functions as a coreceptor expressed on immune cells and facilitates entry of certain HIV-1 virus strains (R5-tropic) By binding CCR5 maraviroc effectively blocks HIV-1 envelope protein gp120 anchoring and consequently prevents viral fusion and subsequent cell entry In cellular assays with CCR5-tropic HIV-1 maraviroc demonstrates substantial antiviral activity (IC50 approximately 2 0 nM) and efficiently inhibits gp120-CCR5 interaction Maraviroc serves as a valuable research tool to explore CCR5-mediated viral entry and HIV tropism dynamics