Learn More
Cole-Parmer Antivibration Marble Slab, 20" L x 13" W x 2" H
Catalog No. NC9579507 Shop All Cole-Parmer Products
Encompass_Preferred
Marble slab reduces vibration and shock ideal for spaces where tables won't fit. Measures 20"L x 13"W x 2"H (50.8 x 33 x 5.1 cm).
Catalog No. NC9579507 Supplier Cole-Parmer Supplier No. 1150010
Marble slab reduces vibration and shock ideal for spaces where tables won't fit. Measures 20"L x 13"W x 2"H (50.8 x 33 x 5.1 cm).