Learn More
Gawet Marble and Granite Vermont Marble™ Slab, 24″ x 24″ x 2″ thick
Catalog No. NC0461927
Encompass_Preferred
The Vermont Marble™ Slab is for use on laboratory furniture.   It substantially reduces vibration and shock where a Vermont Marble™ Balance Table cannot be used.– Standard Matte Machined Finish– Includes set of 4 anti-vibration damping pads
Catalog No. NC0461927 Supplier Gawet Marble and Granite Supplier No. 24X24X2
The Vermont Marble™ Slab is for use on laboratory furniture.   It substantially reduces vibration and shock where a Vermont Marble™ Balance Table cannot be used.– Standard Matte Machined Finish– Includes set of 4 anti-vibration damping pads