Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NoTrax™ Marble Sof-Tyle™ Anti-Fatigue Mat
GREENER_CHOICE

Catalog No. 19015808 Shop All NoTrax Products
Change view
Click to view available options
Dimensions (L x W):
75 x 3 ft.
75 x 4 ft.
Color:
Black Smoke
Blue
Smoke Gray
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Color
19-015-808 75 x 3 ft. Blue
19-015-807 75 x 3 ft. Smoke Gray
19-015-805 75 x 4 ft. Blue
19-015-804 75 x 4 ft. Smoke Gray
19-015-806 75 x 4 ft. Black Smoke
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-015-808 Supplier NoTrax™ Supplier No. 470R0036BU
Add to Cart
Add to Cart

Marble Sof-Tyle™ is an anti-fatigue mat with a heavy-duty vinyl stylish smooth marble-pattern design with a dense sponge backing that utilizes NoTrax™ exclusive UniFusion™ technology. Comes with RedStop™, a non-slip backing that eliminates slipping and sliding of the mat on smooth surfaces.

  • Marble Sof-Tyle™ is an anti-fatigue floor mat provides comfort and ergonomic support, while also complementing any décor with a stylish smooth marble-pattern design
  • The durable vinyl top surface is easy to sweep clean or damp mop and is combined with a dense closed-cell foam base that utilizes NoTrax™ exclusive UniFusion™ technology which virtually eliminates the possibility of de-lamination
  • NoTrax™ laminate floor mats that come with RedStop™, a uniquely engineered non-slip backing technology, eliminate slipping and sliding of floor mats on smooth surface flooring such as tile, wood, marble, and treated concrete
  • For dry applications such as lab testing area, check-out counters, order fulfillment lines and pharmacies
  • The sloped beveled edges reduce tripping hazards and allow easy access on and off the mat

Specifications

Certifications/Compliance Certified by the NFSI (National Floor Safety Institute)
For Use With (Application) Placement in manufacturing plants, assembly lines, picking, packing and logistics warehouses, individual work stations
Color Blue
Material Nitricell, PVC
Dimensions (L x W) 75 x 3 ft.
Type Anti-fatigue Floor Mat
Thickness (English) 1/2 in.
Thickness (Metric) 0.0127 m
Length (English) 75 ft.
Length (Metric) 22.86 m
Width (English) 3 ft.
Width (Metric) 0.9144 m
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.