Learn More
Medchemexpress LLC Marbofloxacin | 115550-35-1 | 99.8% | 100 MG
SDP
Catalog No. 502082372
Encompass_Preferred
Marbofloxacin is a third-generation fluoroquinolone and an orally active antimicrobial agent. It possesses broad-spectrum bactericidal activity and good efficacy, making it suitable for research into infections caused by Gram-positive and Gram-negative bacteria and Mycoplasma.
  • Third-generation fluoroquinolone
  • Orally active antimicrobial agent
  • Exhibits broad-spectrum bactericidal activity
  • Effective against Gram-positive and Gram-negative bacteria
  • Effective against Mycoplasma
  • High purity of 99.8%
  • Appears as a light yellow to yellow solid
Catalog No. 50-208-2372 Supplier Medchemexpress LLC Supplier No. HYB0126100MG
Marbofloxacin is a third-generation fluoroquinolone and an orally active antimicrobial agent. It possesses broad-spectrum bactericidal activity and good efficacy, making it suitable for research into infections caused by Gram-positive and Gram-negative bacteria and Mycoplasma.
  • Third-generation fluoroquinolone
  • Orally active antimicrobial agent
  • Exhibits broad-spectrum bactericidal activity
  • Effective against Gram-positive and Gram-negative bacteria
  • Effective against Mycoplasma
  • High purity of 99.8%
  • Appears as a light yellow to yellow solid