Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marbofloxacin, G-Biosciences
SCI_ED

Catalog No. S139760
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139760 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139760 Supplier G-Biosciences Supplier No. RC1048
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Marbofloxacin is a third generation fluoroquinolone antibiotic used to target Pseudomonades and Staphylococci species.

It also targets a variety of other gram-positive and gram-negative bacteria.

Chemical Identifiers

Molecular Weight (g/mol) 362.36

Specifications

Physical Form Powder
Quantity 1 g
Product Line Gibco
Boiling Point 570.5°C
Percent Purity 0.98
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.