Learn More
Immunology Consultants Purified Marburg Virus Nucleoprotein - Recombinant, 0.1 mg
SDP
Catalog No. NC2734290
Encompass_Preferred
Recombinant Marburg Virus Nucleoprotein with a 6xHIS tag was expressed in CHO cells and purified by nickel affinity chromatography.  Estimated purity is 90%.
Catalog No. NC2734290 Supplier Immunology Consultants Supplier No. AG55022502Z
Recombinant Marburg Virus Nucleoprotein with a 6xHIS tag was expressed in CHO cells and purified by nickel affinity chromatography.  Estimated purity is 90%.