Learn More
Immunology Consultants Purified Marburg Virus VP40 - Recombinant, 0.1 mg
SDP
Catalog No. NC2734292
Encompass_Preferred
Recombinant Marburg Virus VP40 (UniProt # P35260) with a 6xHIS tag was expressed in CHO cells and purified by nickel affinity chromatography.  Estimated purity is >95%.
Catalog No. NC2734292 Supplier Immunology Consultants Supplier No. AG55022601Z
Recombinant Marburg Virus VP40 (UniProt # P35260) with a 6xHIS tag was expressed in CHO cells and purified by nickel affinity chromatography.  Estimated purity is >95%.