Learn More
MSC Mahr MarCal 16 ER Digital Caliper, Hardened Stainless Steel, 150 mm Range, 1 ea
Catalog No. 5000516041
Encompass
The MarCal 16 ER digital caliper is a precision measurement tool designed for versatile inside, outside, depth, and step dimensioning. Constructed from hardened stainless steel, this instrument features a reference system that allows for immediate measurements upon activation. The high-contrast display and integrated features ensure reliable operation in various technical applications.
  • Hardened stainless steel slide and beam for durability
  • Immediate measurement capability via reference system
  • Integrated dirt wipers in the slide for protection
  • High-contrast digit and analog display
  • Includes depth measurement rod and inside diameter jaws
  • Step measuring function
  • Raised guideway for scale protection
  • Operates on a CR 2032 3V lithium battery
  • Includes case and instruction manual
Catalog No. 50-005-16041 Supplier MSC Supplier No. 66867102
The MarCal 16 ER digital caliper is a precision measurement tool designed for versatile inside, outside, depth, and step dimensioning. Constructed from hardened stainless steel, this instrument features a reference system that allows for immediate measurements upon activation. The high-contrast display and integrated features ensure reliable operation in various technical applications.
  • Hardened stainless steel slide and beam for durability
  • Immediate measurement capability via reference system
  • Integrated dirt wipers in the slide for protection
  • High-contrast digit and analog display
  • Includes depth measurement rod and inside diameter jaws
  • Step measuring function
  • Raised guideway for scale protection
  • Operates on a CR 2032 3V lithium battery
  • Includes case and instruction manual