Learn More
Aviva Systems Biology Corp MARCH2 Antibody - middle region (ARP43219_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43219_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against March2. It was validated on Western Blot. . Alternate names: 9530046H09Rik, MGC7259, MARCH-IIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-063